Acros Organics Bromocresol Green, reagent ACS (10g) CAS 76-60-8

Representative image only.

Acros Organics Bromocresol Green, reagent ACS (10g) CAS 76-60-8

Acros Organics | Mfr: AC402930100| Item: UX-88340-62

Reagents and solutions to meet your lab and research needs

$133.25USD / Each

$133.25USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaC21H14Br4O5S
  • ClassSolids
  • ChemistryAnalytical Reagents
  • FormPowder
  • Synonyms3',3',5',5'-Tetrabromo-m-cresolsulfonephthalein, BCG
  • Weight (g)10
  • Formula Weight698.02
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • CAS Number76-60-8
  • DescriptionBromocresol Green, reagent ACS (10g) CAS 76-60-8
Documents & Certificates
Reviews
Q&A