Acros Organics Bromocresol Purple, water soluble, indicator grade (5g)

Representative image only.

Acros Organics Bromocresol Purple, water soluble, indicator grade (5g)

Acros Organics | Mfr: AC229610050| Item: UX-88246-18

Quality and consistency your lab can count on

$76.00USD / Each

$76.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeIndicator
  • Empirical FormulaC21H15Br2NaO5S
  • ClassSolids
  • ChemistrySolutions
  • Synonyms5',5'-Dibromo-o-cresolsulfonephthalein, sodium salt, BCP, Bromcresol Purple, sodium salt
  • FormPowder
  • Weight (g)5
  • Formula Weight562.2
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • CAS Number62625-30-3
  • DescriptionBromocresol Purple, water soluble, indicator grade (5g)
Documents & Certificates
Reviews
Q&A