Acros Organics Lactic acid, reagent ACS (500g) CAS 50-21-5

Representative image only.

Acros Organics Lactic acid, reagent ACS (500g) CAS 50-21-5

Acros Organics | Mfr: AC412965000| Item: UX-88350-08

Quality and consistency your lab can count on

$156.50USD / Each

$156.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Irritant
Specifications
  • Empirical FormulaC3H6O3
  • Linear FormulaCH3CHOHCOOH
  • ClassReagent Acids and Bases
  • SynonymsDL-2-Hydroxypropanoic acid
  • FormSolid
  • Weight (g)500
  • Formula Weight90.08
  • Hazard SymbolIrritant
  • CAS Number50-21-5
  • DescriptionLactic acid, reagent ACS (500g) CAS 50-21-5
Documents & Certificates
Reviews
Q&A