Airfiltronix FR-2 Activated Charcoal Filter, 1.5" Deep; 3/Box
Representative image only.
Airfiltronix FR-2 Activated Charcoal Filter, 1.5" Deep; 3/Box
Airfiltronix | Mfr: FR-2| Item: UX-33705-60
A filter for every application
$334.00USD / Box of 3
$334.00USD / Box of 3
Buy It Together
Current Item
Airfiltronix FR-2 Activated Charcoal Filter, 1.5" Deep; 3/Box
Airfiltronix FR-2 Activated Charcoal Filter, 1.5" Deep; 3/Box
3370560
$334.00USD / Box of 3
+ + + + + =
Total Price 6303.50 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
  • Adsorbs most commonly found organic vapors and odors
  • Good for most organic solvent fumes
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
Specifications
  • Length (InchShort)10 1/2
  • Width (InchShort)10 1/2
  • Depth (InchShort)1 1/2
  • DescriptionActivated Charcoal Filter, 1.5" Deep; 3/Box