Ansell 92-500 585190 Nitrile Gloves, Powdered, Large
Representative image only.
Ansell 92-500 585190 Nitrile Gloves, Powdered, Large
Ansell | Mfr: 92-500 585190| Item: UX-81602-60
Standard in comfort, durability, and cleanliness
$29.70USD / Box of 100
BUY MORE & SAVE
  • 1-9$29.70 USD / Box of 100
  • 10+$26.73 USD / Box of 100
$29.70USD / Box of 100
BUY MORE & SAVE
  • 1-9$29.70 USD / Box of 100
  • 10+$26.73 USD / Box of 100
Product Description
Key Features
  • Resist punctures and tear
  • Ambidextrous
More About this Item
Premium quality gloves are thin enough to allow excellent dexterity, yet tough enough to resist punctures and tears. Gloves are 100% nitrile and contain no fillers, silicones, or plasticizers.
Specifications
  • Thickness (mil)4
  • MaterialNitrile
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • ComplianceFDA-accepted
  • SterileNo
  • PowderedYes
  • DescriptionNitrile Gloves, Powdered, Large