Aven Tools Non-Serrated Color Coded Tweezer Set; 6 Piece Set
Representative image only.
Aven Tools Non-Serrated Color Coded Tweezer Set; 6 Piece Set
Aven Tools | Mfr: 18480EZF| Item: UX-07291-00
Each style has a unique color for easy identification
$59.30USD / Each
$59.30USD / Each
Product Description
Key Features
  • Anti-acid
  • Anti-magnetic
More About this Item
Durable colored coating for high visibility, helps you quickly identify the tweezer needed for a particular application.
Anti-acid, anti-magnetic stainless steel tweezers are available individually or in a complete set.
Set contains one of each of the following:
Black- Style 2A straight, flat tips
Light Blue - Style 3C straight, ultra-fine tips
Red - Style OO tapered, strong tips
Yellow - Style AA straight, fine tips
Green - Style 5 taperd, extra-fine, pointed tips
Orange - Style 7 curved, super-fine, pointed tips
Specifications
  • Overall Length (in)5 3/4
  • MaterialStainless Steel
  • SterileNo
  • ColorAssorted
  • StyleAssorted Tips
  • DescriptionNon-Serrated Color Coded Tweezer Set; 6 Piece Set