Chemetrics K-8012 Phenol CHEMets Visual Kit

Representative image only.

Chemetrics K-8012 Phenol CHEMets Visual Kit

Chemetrics | Mfr: K-8012| Item: UX-00290-30

CHEMets test kits are portable and self-contained

$103.25USD / Each

$103.25USD / Each

Buy It Together

Current Item

Chemetrics K-8012 Phenol CHEMets Visual Kit
Chemetrics K-8012 Phenol CHEMets Visual Kit
0029030

$103.25USD / Each
+ + +
Chemetrics R-8012 Refill Kit , PHenol Test, for 00290-30
0029032

$50.85USD / Box of 30
+
LaMotte Insta-Test Ammonia Test Strips, 0 to 6 ppm, 25/Pk
9953264

$38.25USD / Pkg of 25
=
Total Price 337.10 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, 25 mL sample cup and instructions.

Key Features

  • Vacuum sealed reagents and handy comparator chart for quick, visual results

More About this Item

CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.

Specifications
  • TestPhenol
  • Measurement Points0 to 1 ppm; 0 to 12 ppm
  • Tests Per Kit30
  • DescriptionPhenol CHEMets Visual Kit
Reviews
Q&A