Cole-Parmer 0.01 to 20 Torr Pressure/Vacuum Controller for Pirani-Type Sensor
Representative image only.
Cole-Parmer 0.01 to 20 Torr Pressure/Vacuum Controller for Pirani-Type Sensor
Cole-Parmer | Item: UX-68801-22
Customize to your needs
$1,040.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,040.00USD / Each
Buy It Together
Current Item
Cole-Parmer 0.01 to 20 torr Pressure/Vacuum Controller for Pirani-Type Sensor
Cole-Parmer 0.01 to 20 torr Pressure/Vacuum Controller for Pirani-Type Sensor
6880122
$1,040.00USD / Each
+ + +
Cole-Parmer Vacuum Sensor Extension Cable, 20'
6880175
$119.25USD / Each
=
Total Price 2405.25 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
  • 2 year warranty
More About this Item
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.
Two year warranty
Specifications
  • Accuracy±5% of reading or ±0.03 Torr
  • Unit Of MeasurementTorr
  • Display3.5-digit LCD
  • Battery Type115 VAC, 50/60 Hz
  • Electrical Connections5.5-ft (1.7-m) power cord
  • Switch output3 A at 115 VAC (noninductive)
  • Output1 mV/digit, SPDT
  • Resolution0.01 Torr
  • Description0.01 to 20 torr Pressure/Vacuum Controller for Pirani-Type Sensor