Cole-Parmer Acetonitrile, HPLC Grade, 1 L

Representative image only.

Cole-Parmer Acetonitrile, HPLC Grade, 1 L

Cole-Parmer | Item: UX-88405-44

Reagents and solutions to meet your lab and research needs

$284.00USD / Each

$284.00USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeHPLC
  • Empirical FormulaC2H3N
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight41.04
  • PackagingAmber Glass Bottle
  • DOT InformationAcetonitrile; Class 3, Packing Group II
  • UN NumberUN1648
  • CAS Number75-05-8
  • DescriptionAcetonitrile, HPLC Grade, 1 L
Reviews
Q&A