Cole-Parmer Ammonium Hydroxide Solution, ACS Grade, 500 mL

Representative image only.

Cole-Parmer Ammonium Hydroxide Solution, ACS Grade, 500 mL

Cole-Parmer | Item: UX-88401-68

Reagents and solutions to meet your lab and research needs

$77.30USD / Each

$77.30USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.28
  • Purity GradeACS Reagent
  • Empirical FormulaNH4OH
  • ClassReagent Base
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • Formula Weight35.05
  • PackagingClear Glass
  • DOT InformationAmmonia solution; Class 8, Packing Group III
  • UN NumberUN2672
  • CAS Number1336-21-6
  • Shelf Life (months)24
  • DescriptionAmmonium Hydroxide Solution, ACS Grade, 500 mL
Reviews
Q&A