Cole-Parmer Dissecting Blade Remover for Dissecting Blade Handles
Representative image only.
Cole-Parmer Dissecting Blade Remover for Dissecting Blade Handles
Cole-Parmer | Item: UX-10822-35
$25.70USD / Each
$25.70USD / Each
Buy It Together
Current Item
Cole-Parmer Dissecting Blade Remover for Dissecting Blade Handles
Cole-Parmer Dissecting Blade Remover for Dissecting Blade Handles
1082235
$25.70USD / Each
+ =
Total Price 104.30 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Choose from Carbon Steel (CS) or Stainless Steel (SS) scalpel blades.
Use with all disposable blades on #3, #4, #7, #8 and Siegel handles.
Note: Not intended for medical applications or patient use.
Specifications
  • MaterialStainless Steel
  • DescriptionDissecting Blade Remover for Dissecting Blade Handles