Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.2 um, 5/pk
Representative image only.
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.2 um, 5/pk
Cole-Parmer | Item: UX-25200-87
Optimize performance of your Cole-Parmer pipettes
$51.80USD / Pkg of 5
$51.80USD / Pkg of 5
Buy It Together
Current Item
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.2 um, 5/pk
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.2 um, 5/pk
2520087
$51.80USD / Pkg of 5
+ =
Total Price 464.80 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your pipette running smoothly with these filters and stands
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your pipette controller.
Chemical-resistant PTFE filters help prevent volatile fumes and accidental liquid overfill from corroding your pipette controller's internal components.
0.2 µm filter also helps to prevent bacterial contamination.
1 Year Warranty
Specifications
  • Compatible With25200-15
  • DescriptionFilter for Comfort-Grip Motorized Pipette Controller, 0.2 um, 5/pk