Cole-Parmer Hexanes, ACS Grade, 4 L

Representative image only.

Cole-Parmer Hexanes, ACS Grade, 4 L

Cole-Parmer | Item: UX-88405-64

Reagents and solutions to meet your lab and research needs

$282.80USD / Each

$282.80USD / Each

Buy It Together

Current Item

Cole-Parmer Hexanes, ACS Grade, 4 L
Cole-Parmer Hexanes, ACS Grade, 4 L
8840564

$282.80USD / Each
+ =
Total Price 837.30 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaC6H14
  • ChemistrySolvents
  • FormLiquid
  • SynonymsNaphtha Solvent, n-Hexane
  • Volume (mL)4000
  • Formula Weight86.17
  • PackagingAmber Glass Bottle
  • DOT InformationHexanes; Class 3, Packing Group II
  • Hazard SymbolFlammable
  • CAS Number110-54-3
  • UN NumberUN1208
  • Shelf Life (days)60
  • DescriptionHexanes, ACS Grade, 4 L
Reviews
Q&A