Cole-Parmer Laboratory Mixer H-Stand; Adjustable to 39 3/4"
Representative image only.
Cole-Parmer Laboratory Mixer H-Stand; Adjustable to 39 3/4"
Cole-Parmer | Mfr: 5070394| Item: UX-50703-94
Keep your overhead mixer running smoothly
$546.00USD / Each
$546.00USD / Each
Buy It Together
Current Item
Cole-Parmer Laboratory Mixer H-Stand; Adjustable to 39 3/4"
Cole-Parmer Laboratory Mixer H-Stand; Adjustable to 39 3/4"
5070394
$546.00USD / Each
+ + + +
Cole-Parmer Bracket Assembly, Adjustable Tilt
5500811
$87.00USD / Each
+ +
Cole-Parmer   GP Mixer Set; 120/230 VAC
5500800
$1,204.00USD / Each
$1,505.00USD / Each
+ + +
Cole-Parmer Heavy-Duty Stand for Overhead Mixer
9996821
$1,440.00USD / Each
=
Total Price 5147.95 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Update clamps, mounts, shafts and impellers to accommodate new protocols. These replacement parts will keep your overhead mixer running smoothly.
Particularly stable stand with H-shape base which prevents the stand from tipping backwards. Additionally equipped with a pneumatic spring stand rod, which enables heavy instruments and attachments to be raised and lowered smoothly. The stand has an adjustable screw which can be used to compensate for an uneven laboratory table surface.
Diameter of rod support: 1 1/3" (34mm)
Height: 24 2/5" to 39 3/4" (620-1010mm)
Two year warranty
Specifications
  • Shaft Diameter (in)1.33
  • Shaft Diameter (mm)34
  • Capacity10kg
  • DescriptionMixer H-stand, adjustable to 39 3/4"