Cole-Parmer Mixing Bowl, 304 SS, 3 Qt Capacity
Representative image only.
Cole-Parmer Mixing Bowl, 304 SS, 3 Qt Capacity
Cole-Parmer | Item: UX-07301-04
304 stainless steel composition for increased heat resistance
$32.10USD / Each
BUY MORE & SAVE
  • 1-5$32.10 USD / Each
  • 6-29$28.89 USD / Each
  • 30-119$27.29 USD / Each
  • 120+$25.68 USD / Each
$32.10USD / Each
BUY MORE & SAVE
  • 1-5$32.10 USD / Each
  • 6-29$28.89 USD / Each
  • 30-119$27.29 USD / Each
  • 120+$25.68 USD / Each
Product Description
Key Features
  • Rounded corners for easy mixing and cleaning
  • Withstand temperatures up to 1472 °F (800 °C)
  • 1 year warranty
1 Year WarrantyNSF international certified
Specifications
  • Capacity (L)2.8
  • Capacity (Qt)3
  • Material304 stainless steel
  • Overall Height (in)4
  • Overall Height (cm)10.2
  • Outside Diameter (OD) (in)9 3/8
  • Outside Diameter (OD) (cm)23.8
  • DescriptionMixing Bowl, 304 SS, 3 Qt Capacity; each