Environmental Express pH flow-through cell, 800 uL, 3 mm ID, glass
Representative image only.
Environmental Express pH flow-through cell, 800 uL, 3 mm ID, glass
Environmental Express | Item: EW-05662-48
Ideal for fermentation applications
$192.00USD / Each
$192.00USD / Each
Buy It Together
Current Item
Environmental Express pH flow-through cell, 800 uL, 3 mm ID, glass
Environmental Express pH flow-through cell, 800 uL, 3 mm ID, glass
0566248
$192.00USD / Each
+ =
Total Price 299.65 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
  • 1 year warranty
More About this Item
Measure the pH of high-purity water without the problems usually associated with low-ionic strength solutions.
Not a complete system (electrode and cable needed). See accessories section for required extra components.
1 Year Warranty
Specifications
  • Electrode Body MaterialGlass
  • DescriptionpH flow-through cell, 800 uL, 3 mm ID, glass