Cole-Parmer Silver Nitrate, 0.100N (N/10), 500 mL

Representative image only.

Cole-Parmer Silver Nitrate, 0.100N (N/10), 500 mL

Cole-Parmer | Item: UX-88400-69

Reagents and solutions to meet your lab and research needs

$129.50USD / Each

$129.50USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.1
  • Purity GradeComponents all ACS
  • Empirical FormulaAgNO3
  • ClassTitrants
  • ChemistrySolutions
  • SynonymsNitric acid, silver salt
  • FormLiquid
  • Volume (mL)500
  • Formula Weight169.87
  • PackagingAmber Poly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7761-88-8
  • Shelf Life (months)24
  • DescriptionSilver Nitrate, 0.100N (N/10), 500 mL
Reviews
Q&A