Cole-Parmer Sodium Nitrite, ACS Grade, 500 g

Representative image only.

Cole-Parmer Sodium Nitrite, ACS Grade, 500 g

Cole-Parmer | Item: UX-88404-85

Quality and consistency your lab can count on

$83.10USD / Each

$83.10USD / Each

Buy It Together

Current Item

Cole-Parmer Sodium Nitrite, ACS Grade, 500 g
Cole-Parmer Sodium Nitrite, ACS Grade, 500 g
8840485

$83.10USD / Each
+
Reagents Sodium Nitrite, Purified; 500 g
7891398

$49.40USD / Each
+
Reagents Sodium Nitrite, 500g, ACS Reagent
7891399

$202.50USD / Each
=
Total Price 335.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaNaNO2
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight68.99
  • PackagingPoly Bottle
  • DOT InformationSodium Nitrite; Class 5.1, Packing Group III
  • CAS Number7632-00-0
  • UN NumberUN1500
  • Shelf Life (days)60
  • DescriptionSodium Nitrite, ACS Grade, 500 g
Reviews
Q&A