Cole-Parmer Watch Glass 300mm 12/pk
Representative image only.
Cole-Parmer Watch Glass 300mm 12/pk
Cole-Parmer | Item: UX-34508-68
$324.00USD / Pkg of 12
BUY MORE & SAVE
  • 1-4$324.00 USD / Pkg of 12
  • 5-9$291.60 USD / Pkg of 12
  • 10+$275.40 USD / Pkg of 12
$324.00USD / Pkg of 12
BUY MORE & SAVE
  • 1-4$324.00 USD / Pkg of 12
  • 5-9$291.60 USD / Pkg of 12
  • 10+$275.40 USD / Pkg of 12
Product Description
Key Features
  • 1 year warranty
More About this Item
Manufactured from clear glass sheet (1.8 to 2.0mm thick), these watch glasses are fire ground and have polished edges. Concave shape prevents sliding and return of condensation to the beaker.
1 Year Warranty
Specifications
  • Fits Bottle / Beaker Size (mL)11000
  • MaterialGlass
  • AutoclavableYes
  • Outside Diameter (OD) (cm)30
  • DescriptionWatch Glass 300mm 12/pk