D/E neutralizing broth, prepared; 750 mL bottle, 10/pk

Representative image only.

D/E neutralizing broth, prepared; 750 mL bottle, 10/pk

Hardy Diagnostics| Item: UX-14204-58

Save time and money with ready-made bottles

$429.00USD / Pkg of 10

$429.00USD / Pkg of 10
Product Description

Key Features

  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For neutralizing and evaluating the efficacy of antiseptics and disinfectants
  • 6 month warranty

More About this Item

Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

6 Month Warranty
Specifications
  • Media Name (No.)D/E Neutralizing
  • Capacity (mL)750
  • Capacity (L)0.75
  • DescriptionD/E neutralizing broth, prepared; 750 mL bottle, 10/pk
  • Warranty6 month
Reviews
Q&A