Drager 8318785 Mulitgas Monitor Battery Kit; 110V

Representative image only.

Drager 8318785 Mulitgas Monitor Battery Kit; 110V

Drager | Mfr: 8318785| Item: UX-86515-23

Optimize performance of your air/gas analyzer

$467.00USD / Each

$467.00USD / Each
Product Description

Includes

Cradle and power supply.

Key Features

  • Compatible with Drager multigas detector models 86515-20 through -22
  • Nickel-Metal Hydride battery type
  • Battery life of up to 12 hours
  • 1 year warranty

More About this Item

Battery kit, bump test station, and other accessories to customize your sampling and analysis system.

Multigas detector sold separately.

1 Year Warranty
Specifications
  • Compatible WithDräger X-am® 2000
  • Battery1 NiMH (included)
  • Power (VAC)110
  • DescriptionMulitgas Monitor Battery Kit; 110V
  • Warranty1 year
Reviews
Q&A