Edwards A65201131 Clean and Overhaul Kit for RV-Series Oil-Sealed Rotary Vane Pumps
Representative image only.
Edwards A65201131 Clean and Overhaul Kit for RV-Series Oil-Sealed Rotary Vane Pumps
Edwards | Mfr: A65201131| Item: EW-79303-02
Repair kits help prolong the life of your vacuum pump
$849.00USD / Each
$849.00USD / Each
Product Description
Includes
1x oldham coupling RV3/5/8/12, 1x exhaust diaphragm FKM GP RVP, 1x U seal FKM GP RVP, 1x shoulder washer (ballast) RV, 1x spring (ballast control) St/St, 1x outer shaft seal spacer RV, 3x O-ring Nit 21.5 ID x 3.0 Sec, 1x O-ring Nit 49.5 ID x 3.0 sec, 1x O-ring Nit 32.5 ID x 3.0 sec, 1x O-ring FKM GP 7.6 ID x 2.4 sec, 1x spring (inlet valve) St/St RVP, 1x O-ring FKM GP 57.6 ID x 2.4 sec, 1x O-ring Nit 42.5 ID x 3.0 sec, 1x top plate gasket, 1x shaft seal babsl 20 x 30 x 7 RV, 1x oil pressure valve RV, 1x valve pad E2M1.5 E2M18 pump, 2x spring (dump valve) RV, 1x oil box printed gasket RV, 1x printed gasket cartridge RV, 3x O-ring FKM GP 1.109 ID x.139sec, 1x O-ring FKM GP 9.6 ID x 2.4 sec, 1x seal carrier gasket, 1x air bleed assy RV, 1x RV reed valve, 1x lip seal FKM GP 25 35 7, 1x RV dump valve, 1x O-ring FKM GP 69.5 ID x 3.0 sec, and 1x inlet valve pad order kit.
Key Features
  • Keep service parts handy to minimize downtime
  • Service kit for 79300-series (RV) vacuum pumps
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
The clean and overhaul kit contains all of the springs, gaskets, seals, and other components required for routine maintenance and whenever the pump is dismantled.
Fluoroelastomer clean and overhaul kit works with the following models:
RV3 Rotary Vane Pump 61.7 LPM
RV5 Rotary Vane Pump 96.7 LPM
RV8 Rotary Vane Pump 162.7 LPM
RV12 Rotary Vane Pump 237 LPM
E-LAB 2 Rotary Pump 61.7 LPM
1 Year Warranty
Specifications
  • Compatible WithE2M28 oil-sealed rotary vane vacuum pumps
  • TypeClean and Overhaul Kit
  • DescriptionClean and Overhaul Kit for RV-Series Oil-Sealed Rotary Vane Pumps