Edwards Fomblin® YVAC 06/6 Fluid for Oil Sealed Rotary Pumps, 5 kg (2660 mL)
Representative image only.
Edwards Fomblin® YVAC 06/6 Fluid for Oil Sealed Rotary Pumps, 5 kg (2660 mL)
Edwards | Mfr: H11301020| Item: UX-79300-73
Enhance the performance of your vacuum pumps with the right vacuum pump oil
$3,685.00USD / Each
$3,685.00USD / Each
Product Description
Key Features
  • Specifically designed for vacuum pumps and applications
  • Chemically inert perfluoropolyether oil help keep your vacuum pump running at its best
  • 5 kg (2660 ml)
  • 1 year warranty
More About this Item
Select the right pump oil for your vacuum pumps to keep your equipment in excellent working condition.
The YVAC vacuum pump oil is formulated to ensure optimum pump performance and pump life. Chemically inert perfluoropolyether oil ideal for use in sealed rotary vacuum pumps. Use the oil in chemical processes which require inert mechanical vacuum pump oils.
1 Year Warranty
Specifications
  • DescriptionFomblin® YVAC 06/6 fluid 5 kg (2660 ml) for Oil Sealed Rotary Pumps