Environmental Express Amber VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid; 72/Case

Representative image only.

Environmental Express Amber VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid; 72/Case

Environmental Express | Item: UX-35204-95

Minimize hazardous chemical exposure with pre-filled vials

$78.25USD / Case of 72

$78.25USD / Case of 72

Buy It Together

Current Item

Environmental Express Amber VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid; 72/Case
Environmental Express Amber VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid; 72/Case
3520495

$78.25USD / Case of 72
+ + =
Total Price 244.60 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • 3 month warranty
  • Model number APC1628

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

3 Month Warranty
Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Thread24 mm
  • Septum Type0.125" thick standard septa
  • SterileNo
  • AutoclavableNo
  • Height (in)3 7/8
  • Outside Diameter (in)1 1/8
  • Height (mm)98.81
  • Outside Diameter (mm)27.69
  • Fill0.2 mL, 1:1 hydrochloric acid (HCI)
  • Level1
  • Special Features
  • DescriptionAmber VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid; 72/Cs
  • Warranty3 month
Reviews
Q&A