Environmental Express APC1556 Clear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 0.02% Sodium Thiosulfate; 72/Case

Representative image only.

Environmental Express APC1556 Clear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 0.02% Sodium Thiosulfate; 72/Case

Environmental Express | Item: UX-35205-78

Minimize hazardous chemical exposure with pre-filled vials

$75.30USD / Case of 72

$75.30USD / Case of 72

Buy It Together

Current Item

Environmental Express APC1556 Clear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 0.02% Sodium Thiosulfate; 72/Case
Environmental Express APC1556 Clear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 0.02% Sodium Thiosulfate; 72/Case
3520578

$75.30USD / Case of 72
+ + =
Total Price 241.65 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • Model number APC1556

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • SterileNo
  • AutoclavableNo
  • Outside Diameter (in)1 1/8
  • Outside Diameter (mm)27.69
  • Fill0.2 mL 0.02% Sodium Thiosulfate
  • Level1
  • Special Features
  • DescriptionClear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL 0.02% Sodium Thiosulfate; 72/Case
Reviews
Q&A