Environmental Express APC1594 Clear VOA Vials, 40 mL, Open-Top Cap, 0.5 mL Phosphoric acid; 72/Cs

Representative image only.

Environmental Express APC1594 Clear VOA Vials, 40 mL, Open-Top Cap, 0.5 mL Phosphoric acid; 72/Cs

Environmental Express | Item: UX-35205-89

Minimize hazardous chemical exposure with pre-filled vials

$96.75USD / Case of 72

$96.75USD / Case of 72

Buy It Together

Current Item

Environmental Express APC1594 Clear VOA Vials, 40 mL, Open-Top Cap, 0.5 mL Phosphoric acid; 72/Cs
Environmental Express APC1594 Clear VOA Vials, 40 mL, Open-Top Cap, 0.5 mL Phosphoric acid; 72/Cs
3520589

$96.75USD / Case of 72
+ + =
Total Price 263.10 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • Model number APC1594

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • SterileNo
  • AutoclavableNo
  • Outside Diameter (in)1 1/8
  • Outside Diameter (mm)27.69
  • Fill0.5 mL Phosphoric acid
  • Level1
  • Special Features
  • DescriptionClear VOA Vials, 40 mL, Open-Top Cap, 0.5 mL Phosphoric acid; 72/Cs
Reviews
Q&A