Environmental Express BPC1193 Oblong Packer Wide-Mouth Bottle, HDPE, Level 1, 250 mL, 1 mL Sodium hydroxide; 24/CS
Representative image only.
Environmental Express BPC1193 Oblong Packer Wide-Mouth Bottle, HDPE, Level 1, 250 mL, 1 mL Sodium hydroxide; 24/CS
Environmental Express | Item: UX-35207-72
Won't break like glass
$28.45USD / Case of 24
$28.45USD / Case of 24
Product Description
Key Features
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Lightweight design with leakproof cap
  • Constructed of durable HDPE
Specifications
  • Capacity (mL)250
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap / Stopper Size43/400
  • Cap ColorWhite
  • Bottle ColorNatural
  • Level Number1
  • Special Features1 mL Sodium hydroxide
  • Neck Inside Diameter (ID) (mm)43
  • DescriptionOblong Packer Wide-Mouth Bottle, HDPE, Level 1, 250 mL, 1 mL Sodium hydroxide; 24/CS