Environmental Express Clear VOA Vials, 40 mL, Open-Top Cap, 25 mg Ascorbic Acid; 72/Cs

Representative image only.

Environmental Express Clear VOA Vials, 40 mL, Open-Top Cap, 25 mg Ascorbic Acid; 72/Cs

Environmental Express | Item: UX-35207-03

Minimize hazardous chemical exposure with pre-filled vials

$99.95USD / Case of 72

$99.95USD / Case of 72

Buy It Together

Current Item

Environmental Express Clear VOA Vials, 40 mL, Open-Top Cap, 25 mg Ascorbic Acid; 72/Cs
Environmental Express Clear VOA Vials, 40 mL, Open-Top Cap, 25 mg Ascorbic Acid; 72/Cs
3520703

$99.95USD / Case of 72
+ + =
Total Price 266.30 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • 3 month warranty
  • Model number APC1750

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

3 Month Warranty
Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Thread24 mm
  • Septum Type0.125" thick standard septa
  • SterileNo
  • AutoclavableNo
  • Height (in)3 7/8
  • Outside Diameter (in)1 1/8
  • Height (mm)98.81
  • Outside Diameter (mm)27.69
  • Fill25 mg ascorbic acid (C6H8O6)
  • Level1
  • Special Features
  • DescriptionClear VOA Vials, 40 mL, Open-Top Cap, 25 mg Ascorbic Acid; 72/Cs
  • Warranty3 month
Reviews
Q&A