Environmental Express Clear VOA Vials, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid, 40 mL; 72/Cs

Representative image only.

Environmental Express Clear VOA Vials, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid, 40 mL; 72/Cs

Environmental Express | Item: UX-35204-96

Minimize hazardous chemical exposure with pre-filled vials

$80.35USD / Case of 72

$80.35USD / Case of 72

Buy It Together

Current Item

Environmental Express Clear VOA Vials, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid, 40 mL; 72/Cs
Environmental Express Clear VOA Vials, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid, 40 mL; 72/Cs
3520496

$80.35USD / Case of 72
+ + =
Total Price 246.70 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • 3 month warranty
  • Model number APC1598

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

3 Month Warranty
Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Septum Type0.125" thick standard septa
  • SterileNo
  • AutoclavableNo
  • Height (in)3 7/8
  • Outside Diameter (in)1 1/8
  • Height (mm)98.81
  • Outside Diameter (mm)27.69
  • Fill1.0 mL hydrochloric acid (HCI)
  • Level1
  • Special Features
  • DescriptionClear VOA Vials, Open-Top Cap, 0.2 mL 1:1 Hydrochloric Acid, 40 mL; 72/Cs
  • Warranty3 month
Reviews
Q&A