Environmental Express COD Standard, 1000 mg/L (ppm); 250 mL

Representative image only.

Environmental Express COD Standard, 1000 mg/L (ppm); 250 mL

Environmental Express | Item: UX-35200-11

Controls approved for use in both EPA-compliant and mercury-free vials

$38.20USD / Each

$38.20USD / Each
Product Description

Key Features

  • Made from primary standard grade potassium hydrogen phthalate
  • Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
  • Sold as one 250-mL bottle
  • Easy-to-follow dilution directions printed directly on the label

More About this Item

Primary Standard Grade Potassium Hydrogen Phthalate. Refrigeration is recommended.

Specifications
  • Purity GradePrimary Standard
  • Empirical FormulaC8H5KO4
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • SynonymsPotassium Hydrogen Phthalate; KHP
  • Volume (mL)250
  • Weight (mL)250
  • Packaging250 mL Glass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolRefrigerate
  • CAS Number877-24-7 and 7732-18-5
  • Shelf Life (months)8 (refrigeration required)
  • DescriptionCOD Standard, 1000 mg/L (ppm); 250 mL
Documents & Certificates
Reviews
Q&A