Environmental Express Interf. Check Stnd Mix 3 for ICP-MS in 2% HNO3+Tr HF (12 Compts), Soln A; 250 mL

Representative image only.

Environmental Express Interf. Check Stnd Mix 3 for ICP-MS in 2% HNO3+Tr HF (12 Compts), Soln A; 250 mL

Environmental Express | Item: UX-97063-13

Multi-component interference check standards for ICP-MS analysis

$386.00USD / Each

$386.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.8 and 6020B
  • Solution A of Interference Check Standard Mix 3 Set 97063-12
  • Interference check standard mix 3 for ICP-MS in 2% HNO3 + Tr HF
  • 250 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 12 components.

• Interference check standard mix 3 solution A is a 250 mL bottle which contains 18,000 µg/mL (18,000 ppm) of Chloride; 3,000 µg/mL (3,000 ppm) of Calcium; 2,500 µg/mL (2,500 ppm) of Iron, Sodium; 2,000 µg/mL (2,000 ppm) of Carbon; 1,000 µg/mL (1,000 ppm) of Aluminum, Magnesium, Phosphorus, Potassium, Sulfur; and 20 µg/mL (20 ppm) of Molybdenum, Titanium in 2% HNO3 + Tr HF.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix2% HNO3 & Tr HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)250
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionInterference Check Standard Mix 3 for ICP-MS in 2% HNO3 & Tr HF (12 Components), Solution A; 250 mL
Documents & Certificates
Reviews
Q&A