Environmental Express Molybdenum, 1,000 µg/g for ICP in Mineral Oil; 100 g

Representative image only.

Environmental Express Molybdenum, 1,000 µg/g for ICP in Mineral Oil; 100 g

Environmental Express | Item: UX-97066-45

Single-component metallo-organic standards for ICP analysis

$128.00USD / Each

$128.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified and ISO/IEC 17025:2017 accredited
  • For ASTM Methods D7151, D5708, and D5185
  • 1,000 ppm molybdenum metallo-organic standard in mineral oil matrix
  • 100 g bottle

More About this Item

Use the metallo-organic standards for the determination of metals in oils and lubricants.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/g (1,000 ppm)
  • MatrixMineral Oil
  • Element/IonMolybdenum (Mo)
  • FormLiquid
  • Weight (g)100
  • DOT InformationNot Regulated
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • DescriptionMolybdenum, 1,000 µg/g for ICP in Mineral Oil; 100 g
Documents & Certificates
Reviews
Q&A