Environmental Express Nitrite from Sodium Nitrite, 100 µg/mL for IC in H2O; 250 mL

Representative image only.

Environmental Express Nitrite from Sodium Nitrite, 100 µg/mL for IC in H2O; 250 mL

Environmental Express | Item: UX-97017-76

Single-component standards for ion chromatography

$63.00USD / Each

$63.00USD / Each

Buy It Together

Current Item

Environmental Express Nitrite from Sodium Nitrite, 100 µg/mL for IC in H2O; 250 mL
Environmental Express Nitrite from Sodium Nitrite, 100 µg/mL for IC in H2O; 250 mL
9701776

$63.00USD / Each
+ +
Reagents Sodium Nitrite, 500g, ACS Reagent
7891399

$192.75USD / Each
+
Reagents Sodium Nitrite, Purified; 500 g
7891398

$46.60USD / Each
=
Total Price 410.85 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • For US EPA Methods 300.0 and 300.1
  • 100 ppm nitrite anion from sodium nitrite in H2O matrix
  • 250 mL bottle

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixH2O
  • Element/IonNitrite from Sodium Nitrite
  • FormLiquid
  • Volume (mL)250
  • DOT InformationNot Regulated
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • DescriptionNitrite from Sodium Nitrite, 100 µg/mL in H2O; 250 mL
Documents & Certificates
Reviews
Q&A