Reagents Sodium Nitrite, 10% (w/v), 500mL, Volumetric

Representative image only.

Reagents Sodium Nitrite, 10% (w/v), 500mL, Volumetric

Reagents | Mfr: CS149000-500A| Item: UX-78913-97

Manufactured using the highest quality raw materials in ISO 9001 facility

$114.00USD / Each

$114.00USD / Each

Buy It Together

Current Item

Reagents Sodium Nitrite, 10% (w/v), 500mL, Volumetric
Reagents Sodium Nitrite, 10% (w/v), 500mL, Volumetric
7891397

$114.00USD / Each
+ =
Total Price 180.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Aqueous
  • 1 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

1 Year Warranty
Specifications
  • ClassSolution
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • Shelf Life (days)380
  • Shelf Life (months)12
  • DescriptionSodium Nitrite, 10% (w/v), 500mL, Volumetric
  • Warranty1 year
Documents & Certificates
Reviews
Q&A