Environmental Express Trace Metals in Drinking Water Mix B (26 Components) in 2% HNO3 + Tr HF; 500 mL

Representative image only.

Environmental Express Trace Metals in Drinking Water Mix B (26 Components) in 2% HNO3 + Tr HF; 500 mL

Environmental Express | Item: UX-97015-47

Multi-component standards for drinking water analysis

$540.00USD / Each

$540.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to the NIST SRM 3100 series
  • For US EPA Method 200.8
  • Mix B of Trace Metals in Drinking Water
  • 500 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 26 components.

• Trace metals in drinking water standard Mix B is a 500 mL bottle which contains 31,000 µg/L (31 ppm) of Calcium; 22,000 µg/L (22 ppm) of Sodium; 8,000 µg/L (8 ppm) of Magnesium; 2,500 µg/L (2,500 ppb) of Potassium; 500 µg/L (500 ppb) of Barium; 300 µg/L (300 ppb) of Strontium; 150 µg/L (150 ppb) of Boron; 125 µg/L (125 ppb) of Aluminum; 110 µg/L (110 ppm) of Molybdenum; 90 µg/L (90 ppb) of Iron; 75 µg/L (75 ppb) of Zinc; 60 µg/L (60 ppb) of Nickel; 55 µg/L (55 ppb) of Antimony; 40 µg/L (40 ppb) of Manganese; 35 µg/L (35 ppb) of Vanadium; 25 µg/L (25 ppb) of Cobalt; 20 µg/L (20 ppb) of Chromium, Copper, Lead; 15 µg/L (15 ppb) of Beryllium, Lithium; 11 µg/L (11 ppb) of Selenium; 10 µg/L (10 ppb) of Arsenic, Cadmium, Thallium; and 2 µg/L (2 ppb) of Silver in 2% HNO3 + Tr HF.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix2% HNO3 & Tr HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionTrace Metals in Drinking Water Mix B (26 Components) in 2% HNO3 & Tr HF; 500 mL
Documents & Certificates
Reviews
Q&A