Environmental Express US EPA Method 200.8 Calibration Standard for ICP-MS (2 Components) in 2% HNO3 + Tr HF, Solution B; 100 mL

Representative image only.

Environmental Express US EPA Method 200.8 Calibration Standard for ICP-MS (2 Components) in 2% HNO3 + Tr HF, Solution B; 100 mL

Environmental Express | Item: UX-97062-26

Multi-component calibration standards for ICP-MS analysis

$55.90USD / Each

$55.90USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Method 200.8
  • Solution B of US EPA Method 200.8 Calibration Standard Set 97062-24
  • 100 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 2 components.

• US EPA Method 200.8 calibration standard is a 100 mL bottle which contains 20 µg/mL (20 ppm) of Antimony and Molybdenum in 2% HNO3 + Tr HF.

Hazardous material shipping fees may apply.

Specifications
  • Concentration20 µg/mL (20 ppm)
  • Matrix2% HNO3 & Tr HF
  • Element/IonAntimony, Molybdenum
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Hydrofluoric acid, Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionUS EPA Method 200.8 Calibration Standard for ICP-MS (2 Components) in 2% HNO3 & Tr HF, Solution B; 100 mL
Documents & Certificates
Reviews
Q&A