Environmental Express US EPA Method 200.8 Calibration Standard Set for ICP-MS (20 Components) in 2% HNO3 + Tr HF; 100 mL

Representative image only.

Environmental Express US EPA Method 200.8 Calibration Standard Set for ICP-MS (20 Components) in 2% HNO3 + Tr HF; 100 mL

Environmental Express | Item: UX-97062-24

Multi-component calibration standards for ICP-MS analysis

$301.00USD / Each

$301.00USD / Each

Buy It Together

Current Item

Environmental Express US EPA Method 200.8 Calibration Standard Set for ICP-MS (20 Components) in 2% HNO3 + Tr HF; 100 mL
Environmental Express US EPA Method 200.8 Calibration Standard Set for ICP-MS (20 Components) in 2% HNO3 + Tr HF; 100 mL
9706224

$301.00USD / Each
+ =
Total Price 578.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Method 200.8
  • Calibration Standard Set for EPA Method 200.8 contains 2 solutions
  • Two 100 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard set contains 2 solutions.

• Solution A is a 100 mL bottle which contains 100 µg/mL (100 ppm) of Selenium; and 20 µg/mL (20 ppm) of Aluminum, Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Nickel, Silver, Thallium, Thorium, Uranium, Vanadium, Zinc in 2% HNO3. This is available individually as part number 97062-25.

• Solution B is a 100 mL bottle which contains 20 µg/mL (20 ppm) of Antimony and Molybdenum in 2% HNO3 + Tr HF. This is available individually as part number 97062-26.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonTwo bottle set: Solution A and Solution B
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionUS EPA Method 200.8 Calibration Standard Set for ICP-MS (20 Components) in 2% HNO3 & Tr HF; 100 mL
Reviews
Q&A