Environmental Express US EPA Method 6020 Standard for ICP-MS (22 Components) in 4% HNO3 + Tr HF; 500 mL

Representative image only.

Environmental Express US EPA Method 6020 Standard for ICP-MS (22 Components) in 4% HNO3 + Tr HF; 500 mL

Environmental Express | Item: UX-97063-50

Multi-component standards for ICP-MS analysis

$759.00USD / Each

$759.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Method 6020B
  • 500 mL bottle

More About this Item

When diluted 100-fold, the Contract Required Detection Limits (CRDL) of the elements approved for EPA Method 6020 CLP-M are met.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 22 components.

• US EPA Method 6020 standard is a 500 mL bottle which contains 5 µg/mL (5 ppm) of Calcium, Magnesium, Potassium, Sodium; 0.2 µg/mL (200 ppb) of Aluminum, Barium; 0.1 µg/mL (100 ppb) of Iron; 0.06 µg/mL (60 ppb) of Antimony; 0.05 µg/mL (50 ppb) of Cobalt, Vanadium; 0.04 µg/mL (40 ppb) of Nickel; 0.025 µg/mL (25 ppb) of Copper; 0.02 µg/mL (20 ppb) of Zinc; 0.015 µg/mL (15 ppb) of Manganese; 0.01 µg/mL (10 ppb) of Arsenic, Chromium, Silver, Thallium; 0.005 µg/mL (5 ppb) of Beryllium, Cadmium, Lead, Selenium in 4% HNO3 + Tr HF.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix4% HNO3 & Tr HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionUS EPA Method 6020 Standard for ICP-MS (22 Components) in 4% HNO3 & Tr HF; 500 mL
Documents & Certificates
Reviews
Q&A