Environmental Express Wavelength Calibration Solution Standard for ICP (15 Components) in 5% HNO3; 100 mL

Representative image only.

Environmental Express Wavelength Calibration Solution Standard for ICP (15 Components) in 5% HNO3; 100 mL

Environmental Express | Item: UX-97014-01

Multi-component standards for wavelength calibration

$197.00USD / Each

$197.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to the NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010B
  • 100 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 15 components.

• Wavelength calibration standard is a 100 mL bottle which contains 500 µg/mL (500 ppm) of Potassium and 50 µg/mL (50 ppm) of Aluminum, Arsenic, Barium, Cadmium, Cobalt, Chromium, Copper, Manganese, Molybdenum, Nickel, Lead, Selenium, Strontium, Zinc in 5% HNO3.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionWavelength Calibration Solution Standard for ICP (15 Components) in 5% HNO3; 100 mL
Documents & Certificates
Reviews
Q&A