Fisher Chemical Acetonitrile, OPTIMA™ Grade, 1 L

Representative image only.

Fisher Chemical Acetonitrile, OPTIMA™ Grade, 1 L

Fisher Chemical | Mfr: A9961| Item: UX-88000-84

Quality and consistency your lab can count on

$293.50USD / Each

$293.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Color (APHA)—≤5
  • Pesticide Residue Analysis (as Heptachlor Epoxide)—≤10ng/L
  • Optical Absorbance at 190nm—≤1.00
  • at 195nm—≤0.15
  • at 200nm—≤0.07
  • at 205nm—≤0.05
  • at 210nm—≤0.04
  • at 220nm—≤0.02
  • at 254nm—≤0.005
  • Fluorescence Background (as Quinine Sulfate)—≤1ppb
  • Water—≤0.01%
  • Residue after Evaporation—≤0.8ppm
  • LC Gradient Suitability at 254nm and 205nm—To pass test
  • Refractive Index at 25°C—1.3405-1.3425
  • Titratable Acid—≤0.008mEq/g
  • Titratable Base—≤0.0006mEq/g
  • Density at 25°C—0.775-0.780g/mL
  • Packaged under Nitrogen, Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableToxicStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • Empirical FormulaCH3CN
  • ChemistrySolvents
  • FormLiquid
  • SynonymsCyanomethane, Ethanenitrile, Ethyl nitrile, Methyl cyanide, Methanecarbonitrile
  • Volume (mL)1000
  • Formula Weight41.05
  • PackagingAmber Glass Bottle
  • DOT InformationAcetonitrile; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Toxic
  • UN NumberUN1648
  • CAS Number75-05-8
  • Boiling Point C (° C)81
  • Melting Point C (° C)-46
  • DescriptionAcetonitrile, OPTIMA™ Grade, 1 L
Documents & Certificates
Reviews
Q&A