Fisher Chemical Ethyl Acetate, OPTIMA™ Grade, 4 L

Representative image only.

Fisher Chemical Ethyl Acetate, OPTIMA™ Grade, 4 L

Fisher Chemical | Mfr: E1964| Item: UX-88008-36

Quality and consistency your lab can count on

$328.25USD / Each

$328.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Color (APHA)—≤5
  • Pesticide Residue Analysis (as Heptachlor Epoxide)—10ng/L
  • Optical Absorbance at 255nm—≤1.00
  • &tab;at 260nm—≤0.12
  • &tab;at 270nm—≤0.015
  • Fluorescence Background (as Quinine Sulfate)—≤1ppb
  • Water—≤0.02%
  • Residue after Evaporation—≤1ppm
  • Refractive Index at 25°C—1.3680-1.3710
  • Density at 25°C—0.893-0.895g/mL
  • Titratable Acid—≤0.0009mEq/g
  • Substances Darkened by H2SO4—To pass test
  • Packaged under Nitrogen, Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableIrritantStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • Empirical FormulaCH3COOC2H5
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsAcetic acid ethyl ester
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight88.11
  • PackagingAmber Glass Bottle
  • DOT InformationEthyl acetate; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • CAS Number141-78-6
  • UN NumberUN1173
  • Boiling Point C (° C)75
  • Melting Point C (° C)-83.5
  • DescriptionEthyl Acetate, OPTIMA™ Grade, 4 L
Documents & Certificates
Reviews
Q&A