Fisher Chemical Glycerol, Molecular Biology Grade, 4 L

Representative image only.

Fisher Chemical Glycerol, Molecular Biology Grade, 4 L

Fisher Chemical | Mfr: BP2294| Item: UX-88058-47

Quality and consistency your lab can count on

$1,189.00USD / Each

$1,189.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeMolecular Biology
  • Empirical FormulaHOCH2CH(OH)CH2OH
  • ChemistrySolvents
  • FormLiquid
  • SynonymsGlycerine
  • Volume (mL)4000
  • Formula Weight92.09
  • PackagingHDPE Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGrey
  • CAS Number56-81-5
  • Boiling Point C (° C)290
  • Melting Point C (° C)18
  • DescriptionGlycerol, Molecular Biology Grade, 4 L
Documents & Certificates
Reviews
Q&A