Fisher Chemical Heptane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L

Representative image only.

Fisher Chemical Heptane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L

Fisher Chemical | Mfr: H350SK4| Item: UX-88010-96

Quality and consistency your lab can count on

$727.00USD / Each

$727.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥96%
  • Color (APHA)—≤10
  • Fluorescence Background (as Quinine Sulfate)—To pass test
  • Refractive Index at 25°C—1.3845-1.3865
  • Residue after Evaporation—≤5ppm
  • Water—≤0.02%
  • Optical Absorbance at 200nm—≤0.75
  • at 215nm—≤0.20
  • at 254nm—≤0.014

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeHPLC
  • Empirical FormulaCH3(CH2)5CH3
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonymsn-Heptane
  • Volume (mL)4000
  • Formula Weight100.2
  • PackagingSafe-Cote™ Amber Glass Bottle
  • DOT InformationHeptanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • UN NumberUN1206
  • CAS Number142-82-5
  • Boiling Point C (° C)98
  • Melting Point C (° C)-91
  • DescriptionHeptane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A