Fisher Chemical Hexanes, Certified ACS Grade, Glass Bottle, 1 L

Representative image only.

Fisher Chemical Hexanes, Certified ACS Grade, Glass Bottle, 1 L

Fisher Chemical | Mfr: H2921| Item: UX-88011-30

Quality and consistency your lab can count on

$236.50USD / Each

$236.50USD / Each

Buy It Together

Current Item

Fisher Chemical Hexanes, Certified ACS Grade, Glass Bottle, 1 L
Fisher Chemical Hexanes, Certified ACS Grade, Glass Bottle, 1 L
8801130

$236.50USD / Each
+
Cole-Parmer Hexanes, ACS Grade, 4 L
8840564

$282.80USD / Each
=
Total Price 519.30 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (5 Isomers total Hexanes plus Methylcyclopentane) — ≥98.5%
  • Color (APHA)—≤10
  • Density at 25°C—0.653-0.673g/mL
  • Residue after Evaporation—≤0.001%
  • Water-soluble Titratable Acid—≤0.0003 mEq/g
  • Sulfur Compounds (as S)—≤0.005%
  • Thiophene—To pass test

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaC6H14
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsNaphtha Solvent, n-Hexane
  • Volume (mL)1000
  • Formula Weight86.18
  • PackagingGlass Bottle
  • DOT InformationHexanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number92112-69-1
  • UN NumberUN1208
  • Boiling Point C (° C)69
  • Melting Point C (° C)-95
  • DescriptionHexanes, Certified ACS Grade, Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A