Fisher Chemical Hexanes, Certified ACS Grade, Steel Pail, 20 L

Representative image only.

Fisher Chemical Hexanes, Certified ACS Grade, Steel Pail, 20 L

Fisher Chemical | Mfr: H29220| Item: UX-88074-39

Quality and consistency your lab can count on

$1,254.00USD / Each

$1,254.00USD / Each

Buy It Together

Current Item

Fisher Chemical Hexanes, Certified ACS Grade, Steel Pail, 20 L
Fisher Chemical Hexanes, Certified ACS Grade, Steel Pail, 20 L
8807439

$1,254.00USD / Each
+
Cole-Parmer Hexanes, ACS Grade, 4 L
8840564

$292.75USD / Each
=
Total Price 1546.75 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaC6H14
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsNaphtha Solvent, n-Hexane
  • Volume (mL)20000
  • Formula Weight86.18
  • PackagingSteel Pail
  • DOT InformationHexanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number92112-69-1
  • UN NumberUN1208
  • Boiling Point C (° C)69
  • Melting Point C (° C)-95
  • DescriptionHexanes, Certified ACS Grade, Steel Pail, 20 L
Documents & Certificates
Reviews
Q&A