Fisher Chemical Hexanes, OPTIMA™ Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Hexanes, OPTIMA™ Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: H3034| Item: UX-88011-12

Quality and consistency your lab can count on

$395.25USD / Each

$395.25USD / Each

Buy It Together

Current Item

Fisher Chemical Hexanes, OPTIMA™ Grade, Glass Bottle, 4 L
Fisher Chemical Hexanes, OPTIMA™ Grade, Glass Bottle, 4 L
8801112

$395.25USD / Each
+
Cole-Parmer Hexanes, ACS Grade, 4 L
8840564

$292.75USD / Each
=
Total Price 688.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (Hexanes) — ≥99.9%
  • Assay (n-Hexane)—≥85%
  • Color (APHA)—≤5
  • Suitability for GC/MS—To pass test
  • Pesticide Residue Analysis (as Heptachlor Epoxide)—≤10ng/L
  • Fluorescence Background (as Quinine Sulfate)—≤1ppb
  • Water—≤0.01%
  • Residue after Evaporation—≤1ppm
  • Refractive Index at 25°C—1.3740-1.3780
  • Density at 25°C—0.653-0.673g/mL
  • Water-soluble Titratable Acid—≤0.0003mEq/g
  • Sulfur Compounds (as S)—≤0.005%
  • Thiophene—To pass test
  • Optical Absorbance at 195nm—≤1.00
  • at 210nm—≤0.25
  • at 220nm—≤0.07
  • at 254nm—≤0.005

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • Empirical FormulaC6H14
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsNaphtha Solvent, n-Hexane
  • Volume (mL)4000
  • Formula Weight86.18
  • PackagingGlass Bottle
  • DOT InformationHexanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number92112-69-1
  • UN NumberUN1208
  • Boiling Point C (° C)69
  • Melting Point C (° C)-95
  • DescriptionHexanes, OPTIMA™ Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A