Fisher Chemical Hydrogen Peroxide Solution, 3% w/v, USP Grade, 500 mL

Representative image only.

Fisher Chemical Hydrogen Peroxide Solution, 3% w/v, USP Grade, 500 mL

Fisher Chemical | Mfr: H312500| Item: UX-88012-22

Quality and consistency your lab can count on

$212.75USD / Each

$212.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 3.0-3.5%
  • Acidity—To pass test
  • Barium—To pass test
  • Heavy Metals (as Pb)—≤5ppm
  • Preservative—≤0.05%
  • Nonvolatile Residue—≤0.15%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Concentration0.03
  • Purity GradeUSP
  • MatrixWater
  • Empirical FormulaH2O2
  • ClassGeneral Solutions
  • FormLiquid
  • SynonymsHydrogen Dioxide, Hyperoxide
  • Volume (mL)500
  • Formula Weight34.01
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolHarmful
  • CAS Number7722-84-1
  • Boiling Point C (° C)100
  • Melting Point C (° C)0
  • DescriptionHydrogen Peroxide Solution, 3% w/v, USP Grade, 500 mL
Documents & Certificates
Reviews
Q&A