Fisher Chemical Hydrogen Peroxide Solution, 30%, Certified ACS Grade, 4 L

Representative image only.

Fisher Chemical Hydrogen Peroxide Solution, 30%, Certified ACS Grade, 4 L

Fisher Chemical | Mfr: H3254| Item: UX-88012-14

Quality and consistency your lab can count on

$1,461.00USD / Each

$1,461.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 30.0-32.0%
  • Color (APHA)—≤10
  • Residue after Evaporation—≤0.002%
  • Titratable Acid—≤0.0006mEq/g
  • Chloride—≤3ppm
  • Nitrate—≤2ppm
  • Phosphate—≤2ppm
  • Sulfate—≤5ppm
  • Ammonium—≤5ppm
  • Heavy Metals (as Pb)—≤1ppm
  • Iron—≤0.5ppm
  • Copper—≤0.1ppm
  • Nickel—≤0.1ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

OxidizingCorrosive
Specifications
  • Concentration0.3
  • Purity GradeCertified ACS
  • MatrixWater
  • Empirical FormulaH2O2
  • ClassGeneral Solutions
  • SynonymsHydrogen Dioxide, Hyperoxide
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight34.01
  • PackagingPoly Bottle
  • DOT InformationHydrogen Peroxide, Aqueous Solutions; Class 5.1 (8), Packing Group II
  • Storage SymbolYellow
  • Hazard SymbolCorrosive, Oxidizing
  • CAS Number7722-84-1
  • UN NumberUN2014
  • Boiling Point C (° C)108
  • Melting Point C (° C)-33
  • DescriptionHydrogen Peroxide Solution, 30%, Certified ACS Grade, 4 L
Documents & Certificates
Reviews
Q&A