Fisher Chemical Pentane, HPLC Grade, 1 L

Representative image only.

Fisher Chemical Pentane, HPLC Grade, 1 L

Fisher Chemical | Mfr: P3991| Item: UX-88018-40

Quality and consistency your lab can count on

$223.25USD / Each

$223.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99%
  • Color (APHA) — ≤5
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.3540-1.3570
  • Residue after Evaporation — ≤5ppm
  • Water — ≤0.02%
  • Optical Absorbance at 190nm — ≤1.00
  • at 200nm — ≤0.30
  • at 210nm — ≤0.10
  • at 254nm — ≤0.014
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeHPLC
  • Empirical FormulaCH3(CH2)3CH3
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonymsn-Pentane
  • Volume (mL)1000
  • Formula Weight72.15
  • PackagingAmber Glass Bottle
  • DOT InformationPentanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable
  • UN NumberUN1265
  • CAS Number109-66-0
  • Boiling Point C (° C)36
  • Melting Point C (° C)-130
  • DescriptionPentane, HPLC Grade, 1 L
Documents & Certificates
Reviews
Q&A