Fisher Chemical Potassium Phosphate Monobasic, Certified ACS Grade, 500 g

Representative image only.

Fisher Chemical Potassium Phosphate Monobasic, Certified ACS Grade, 500 g

Fisher Chemical | Mfr: P285500| Item: UX-88020-96

Quality and consistency your lab can count on

$269.00USD / Each

$269.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (K2PO4) — ≥99.0%
  • Insoluble Matter, Calcium, and NH4OH Precipitate — ≤0.010%
  • Loss on Drying at 105°C — ≤0.20%
  • pH of a 5% Solution at 25°C — 4.1 to 4.5
  • Chloride — ≤0.001%
  • Nitrogen Compounds (as N) — ≤0.001%
  • Sulfate — ≤0.003%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.002%
  • Sodium — ≤0.005%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaKH2PO4
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • SynonymsPotassium Dihydrogen Phosphate
  • FormSolid
  • Weight (g)500
  • Formula Weight136.09
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7778-77-0
  • Melting Point C (° C)253
  • DescriptionPotassium Phosphate Monobasic, Certified ACS Grade, 500 g
Documents & Certificates
Reviews
Q&A